(S)-(-)-N,α-Dimethylbenzylamine 96%

Supplier: Apollo Scientific

OR951273-1G OR951273-25G OR951273-5G
APOSOR951273-1GEA 21.5 EUR
APOSOR951273-1G APOSOR951273-25G APOSOR951273-5G
(S)-(-)-N,α-Dimethylbenzylamine 96%
(S)-(-)-N,α-Dimethylbenzylamine
Formula: C₉H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD00067113
CAS Number: 19131-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR