2-Cyanobenzoic acid 96%
Supplier: Apollo Scientific
|
Warning
|
OR951322-25G
APOSOR951322-25GEA
183
EUR
APOSOR951322-25G
2-Cyanobenzoic acid 96%
2-Cyanobenzoic acid
Formula:
C₈H₅NO₂ MW: 147,13 g/mol Melting Pt: 212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045796 CAS Number: 3839-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...