5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid 98%

Supplier: Apollo Scientific
Warning

OR951491-1G OR951491-250MG
APOSOR951491-1GEA 26.1 EUR
APOSOR951491-1G APOSOR951491-250MG
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid 98%
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid
Formula: C₇H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11846936
CAS Number: 796729-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR