2,6-Diaminopyrazine 95%

Supplier: Apollo Scientific
Warning

OR951645-10G OR951645-1G OR951645-5G
APOSOR951645-10GEA 982 EUR
APOSOR951645-10G APOSOR951645-1G APOSOR951645-5G
2,6-Diaminopyrazine 95%
2,6-Diaminopyrazine
Formula: C₄H₆N₄
Storage Temperature: Refrigerator
MDL Number: MFCD09907864
CAS Number: 41536-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR