Ethyl-4-chloropyrimidine-5-carboxylate 95%
Supplier: Apollo Scientific
|
Warning
|
OR951693-10G
OR951693-1G
OR951693-5G
APOSOR951693-10GEA
735
EUR
APOSOR951693-10G
APOSOR951693-1G
APOSOR951693-5G
Ethyl-4-chloropyrimidine-5-carboxylate 95%
Ethyl-4-chloropyrimidine-5-carboxylate
Formula:
C₇H₇ClN₂O₂ MW: 186,6 g/mol Boiling Pt: 278 °C/60mmHg Storage Temperature: Ambient |
MDL Number:
MFCD07371659 CAS Number: 41103-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...