5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde 95+%

Supplier: Apollo Scientific
Warning

OR951828-1G OR951828-250MG
APOSOR951828-1GEA 252 EUR
APOSOR951828-1G APOSOR951828-250MG
5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde 95+%
5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde
Formula: C₈H₅BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11226918
CAS Number: 878197-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR