4-Chlorophthalide 96%
Supplier: Apollo Scientific
OR951873-10G
OR951873-1G
OR951873-5G
APOSOR951873-10GEA
339
EUR
APOSOR951873-10G
APOSOR951873-1G
APOSOR951873-5G
4-Chlorophthalide 96%
4-Chlorophthalide
Formula:
C₈H₅ClO₂ MW: 168.5800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01718133 CAS Number: 52010-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...