2-(4-Aminophenyl)-2-methylpropanenitrile 96%

Supplier: Apollo Scientific

OR952433-10G OR952433-1G OR952433-5G
APOSOR952433-10GEA 271 EUR
APOSOR952433-10G APOSOR952433-1G APOSOR952433-5G
2-(4-Aminophenyl)-2-methylpropanenitrile 96%
2-(4-Aminophenyl)-2-methylpropanenitrile
Formula: C₁₀H₁₂N₂
MW: 160,2200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909912
CAS Number: 115279-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR