Ethyl-3-acetylbenzoate 98%
Supplier: Apollo Scientific
Synonyms:
Ethyl 3-acetylbenzoate
OR952486-1G
OR952486-5G
APOSOR952486-1GEA
31.9
EUR
APOSOR952486-1G
APOSOR952486-5G
Ethyl-3-acetylbenzoate 98%
Ethyl-3-acetylbenzoate
Formula:
C₁₁H₁₂O₃ MW: 192.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02261283 CAS Number: 37847-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...