Methyl-2-oxobutanoate 95%

Supplier: Apollo Scientific

OR952993-10G OR952993-25G OR952993-5G OR952993-1G
APOSOR952993-10GEA 0 EUR
APOSOR952993-10G APOSOR952993-25G APOSOR952993-5G APOSOR952993-1G
Methyl-2-oxobutanoate 95%
Methyl-2-oxobutanoate
Formula: C₅H₈O₃
MW: 116,115 Da
Storage Temperature: Ambient
MDL Number: MFCD03093092
CAS Number: 3952-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR