Methyl thiazole-2-carboxylate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 2-thiazolecarboxylate

OR953064-5G
APOSOR953064-5GEA 89.9 EUR
APOSOR953064-5G
Methyl thiazole-2-carboxylate 98%
Methyl thiazole-2-carboxylate
Formula: C₅H₅NO₂S
MW: 143,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18075580
CAS Number: 55842-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR