2,6-Dichloro-α-methylbenzyl alcohol 95%
Supplier: Apollo Scientific
OR953192-1G
OR953192-5G
APOSOR953192-1GEA
61.1
EUR
APOSOR953192-1G
APOSOR953192-5G
2,6-Dichloro-α-methylbenzyl alcohol 95%
2,6-Dichloro-α-methylbenzyl alcohol
Formula:
C₈H₈Cl₂O MW: 191,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016853 CAS Number: 53066-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...