Methyl pyridazine-4-carboxylate 95%
Supplier: Apollo Scientific
OR953239-1G
OR953239-5G
APOSOR953239-1GEA
328
EUR
APOSOR953239-1G
APOSOR953239-5G
Methyl pyridazine-4-carboxylate 95%
Methyl pyridazine-4-carboxylate
Formula:
C₆H₆N₂O₂ MW: 138,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09953488 CAS Number: 34231-77-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...