N-Methylcyclopropanamine hydrochloride 95%

Supplier: Apollo Scientific

OR953254-10G OR953254-1G OR953254-5G
APOSOR953254-10GEA 101 EUR
APOSOR953254-10G APOSOR953254-1G APOSOR953254-5G
N-Methylcyclopropanamine hydrochloride 95%
N-Methylcyclopropanamine hydrochloride
Formula: C₄H₁₀ClN
MW: 107,5800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997746
CAS Number: 67376-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR