(3-Chlorophenyl)acetic acid ethyl ester 98%
Supplier: Apollo Scientific
Synonyms:
Ethyl (3-chlorophenyl)acetate, Ethyl 2-(3-chlorophenyl)acetate
OR953372-25G
APOSOR953372-25GEA
95.7
EUR
APOSOR953372-25G
(3-Chlorophenyl)acetic acid ethyl ester 98%
(3-Chlorophenyl)acetic acid ethyl ester
Formula:
(C₆H₄Cl)CH₂COOCH₂CH₃ MW: 198,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07787498 CAS Number: 14062-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...