Isopropyl-2-pyridyl ketone 98%

Supplier: Apollo Scientific

OR953473-1G OR953473-5G
APOSOR953473-1GEA 129 EUR
APOSOR953473-1G APOSOR953473-5G
Isopropyl-2-pyridyl ketone 98%
Isopropyl-2-pyridyl ketone
Formula: C₉H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00778577
CAS Number: 59576-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR