N-(2-Chlorophenyl)-2-cyanoacetamide 95%

Supplier: Apollo Scientific

OR953558-1G OR953558-5G
APOSOR953558-1GEA 217 EUR
APOSOR953558-1G APOSOR953558-5G
N-(2-Chlorophenyl)-2-cyanoacetamide 95%
N-(2-Chlorophenyl)-2-cyanoacetamide
Formula: C₉H₇ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD00463630
CAS Number: 50982-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR