3-(3-Chloropropylcarbamoyl)phenylboronic acid 98%
Supplier: Apollo Scientific
Synonyms:
(3-((3-Chloropropyl)carbamoyl)phenyl)boronic acid
OR9539-1G
OR9539-5G
APOSOR9539-1GEA
139
EUR
APOSOR9539-1G
APOSOR9539-5G
3-(3-Chloropropylcarbamoyl)phenylboronic acid 98%
3-(3-Chloropropylcarbamoyl)phenylboronic acid
Formula:
C₁₀H₁₃BClNO₃ MW: 241,4800 g/mol Melting Pt: 218…220 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08436014 CAS Number: 874288-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...