5-(Aminomethyl)-N,N-dimethyl-2-pyridinamine 98%
Supplier: Apollo Scientific
OR953983-1G
OR953983-250MG
OR953983-5G
APOSOR953983-1GEA
270
EUR
APOSOR953983-1G
APOSOR953983-250MG
APOSOR953983-5G
5-(Aminomethyl)-N,N-dimethyl-2-pyridinamine 98%
5-(Aminomethyl)-N,N-dimethyl-2-pyridinamine
Formula:
C₈H₁₃N₃ MW: 151,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08272104 CAS Number: 354824-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...