8-Hydroxy-1,2,3,4-tetrahydroquinoline 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,2,3,4-Tetrahydroquinolin-8-ol, 1,2,3,4-Tetrahydro-8-hydroxyquinoline
OR954020-1G
OR954020-25G
OR954020-5G
APOSOR954020-1GEA
109
EUR
APOSOR954020-1G
APOSOR954020-25G
APOSOR954020-5G
8-Hydroxy-1,2,3,4-tetrahydroquinoline 96%
8-Hydroxy-1,2,3,4-tetrahydroquinoline
Formula:
C₉H₁₁NO MW: 149,1900 g/mol Melting Pt: 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD02656029 CAS Number: 6640-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...