5-Bromo-3-pyridineboronic acid 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromopyridine-5-boronic acid, 3-Bromopyridyl-5-boronic acid
OR9542-1G
OR9542-5G
APOSOR9542-1GEA
56.8
EUR
APOSOR9542-1G
APOSOR9542-5G
5-Bromo-3-pyridineboronic acid 96%
5-Bromo-3-pyridineboronic acid
Formula:
C₅H₅BBrNO₂ MW: 201,81 g/mol Melting Pt: 260 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02685634 CAS Number: 452972-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...