4-Bromo-3-methoxybenzoic acid 98%

Supplier: Apollo Scientific
Warning

OR9543-5G OR9543-25G
APOSOR9543-5GEA 36.4 EUR
APOSOR9543-5G APOSOR9543-25G
4-Bromo-3-methoxybenzoic acid 98%
4-Bromo-3-methoxybenzoic acid
Formula: C₈H₇BrO₃
Melting Pt: 221…224 °C
Storage Temperature: Ambient
MDL Number: MFCD08436017
CAS Number: 56256-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR