5-(4-Methylphenyl)-1,3,4-oxadiazol-2-ol 98%
Supplier: Apollo Scientific
OR954504-10G
OR954504-1G
OR954504-5G
APOSOR954504-10GEA
754
EUR
APOSOR954504-10G
APOSOR954504-1G
APOSOR954504-5G
5-(4-Methylphenyl)-1,3,4-oxadiazol-2-ol 98%
5-(4-Methylphenyl)-1,3,4-oxadiazol-2-ol
Formula:
C₉H₈N₂O₂ MW: 176,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11046712 CAS Number: 83725-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...