Acetoacetanilide 98%
Supplier: Apollo Scientific
Synonyms:
3-Oxo-N-phenyl butamide, 3-Oxo-N-phenyl butanamide
OR954560-100G
OR954560-1KG
OR954560-500G
APOSOR954560-100GEA
125
EUR
APOSOR954560-100G
APOSOR954560-1KG
APOSOR954560-500G
Acetoacetanilide 98%
Acetoacetanilide
Formula:
C₁₀H₁₁NO₂ MW: 177,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008780 CAS Number: 102-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...