2-Methylthiobenzamide 98%

Supplier: Apollo Scientific

Synonyms: 2-Methylbenzothioamide, Thio-o-toluamide

OR954583-100G OR954583-25G
APOSOR954583-100GEA 435 EUR
APOSOR954583-100G APOSOR954583-25G
2-Methylthiobenzamide 98%
2-Methylthiobenzamide
Formula: C₈H₉NS
MW: 151,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03945350
CAS Number: 53515-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR