7-Hydroxy-4-methoxymethylcoumarin

Supplier: Apollo Scientific
Warning

OR9546-1G OR9546-5G
APOSOR9546-1GEA 109 EUR
APOSOR9546-1G APOSOR9546-5G
7-Hydroxy-4-methoxymethylcoumarin
7-Hydroxy-4-methoxymethylcoumarin
Formula: C₁₁H₁₀O₄
MW: 206,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052607
CAS Number: 157101-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR