3-Aminopyridine 99%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Pyridin-3-amine, 3-Pyridylamine
OR9548-100G
OR9548-250G
APOSOR9548-100GEA
29
EUR
APOSOR9548-100G
APOSOR9548-250G
3-Aminopyridine 99%
3-Aminopyridine
Formula:
C₅H₆N₂ MW: 94,12 g/mol Boiling Pt: 250…252 °C Melting Pt: 57…60 °C Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006400 CAS Number: 462-08-8 EINECS: 207-322-2 UN: 2671 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...