Methyl 5-Chlorothiophene-2-carboxylate 98%
Supplier: Apollo Scientific
Synonyms:
5-Chlorothiophene-2-carboxylic acid methyl ester, Methyl 5-chlorothiophene-2-carboxylate
OR955468-1G
OR955468-5G
APOSOR955468-1GEA
270
EUR
APOSOR955468-1G
APOSOR955468-5G
Methyl 5-Chlorothiophene-2-carboxylate 98%
Methyl 5-Chlorothiophene-2-carboxylate
Formula:
C₆H₅ClO₂S MW: 176,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06203698 CAS Number: 35475-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...