Methyl-2-(1,2-benzisoxazol-3-yl)acetate 95+%
Supplier: Apollo Scientific
|
Warning
|
OR955617-1G
OR955617-5G
APOSOR955617-1GEA
270
EUR
APOSOR955617-1G
APOSOR955617-5G
Methyl-2-(1,2-benzisoxazol-3-yl)acetate 95+%
Methyl-2-(1,2-benzisoxazol-3-yl)acetate
Formula:
C₁₀H₉NO₃ MW: 191,19 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD04117784 CAS Number: 59899-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...