4-Chloro-2-nitrotoluene 98%
Supplier: Apollo Scientific
OR956302-100G
OR956302-1KG
OR956302-500G
APOSOR956302-100GEA
125
EUR
APOSOR956302-100G
APOSOR956302-1KG
APOSOR956302-500G
4-Chloro-2-nitrotoluene 98%
4-Chloro-2-nitrotoluene
Formula:
C₇H₆ClNO₂ MW: 171,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007215 CAS Number: 89-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...