4-Chloro-3-methylbenzaldehyde 95%
Supplier: Apollo Scientific
OR957014-1G
OR957014-5G
APOSOR957014-1GEA
60.9
EUR
APOSOR957014-1G
APOSOR957014-5G
4-Chloro-3-methylbenzaldehyde 95%
4-Chloro-3-methylbenzaldehyde
Formula:
C₈H₇ClO Storage Temperature: Ambient |
MDL Number:
MFCD02261736 CAS Number: 101349-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...