3-Bromo-5-chloropyridine 97%

Supplier: Apollo Scientific
Danger

OR9579-25G OR9579-1G OR9579-5G
APOSOR9579-25GEA 81.5 EUR
APOSOR9579-25G APOSOR9579-1G APOSOR9579-5G
3-Bromo-5-chloropyridine 97%
3-Bromo-5-chloropyridine
Formula: C₅H₃BrClN
MW: 192,44 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD04114221
CAS Number: 73583-39-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR