2,3,5-Collidine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3,5-Trimethylpyridine
OR9582-500G
APOSOR9582-500GEA
197
EUR
APOSOR9582-500G
2,3,5-Collidine 98%
2,3,5-Collidine
Formula:
C₈H₁₁N MW: 121,18 g/mol Boiling Pt: 182…184 °C Density: 0,935 g/cm³ (25 °C) Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00126840 CAS Number: 695-98-7 EINECS: 211-786-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...