cis-3-Carbomethoxycyclopentane-1-carboxylic acid 95%
Supplier: Apollo Scientific
OR958338-1G
OR958338-250MG
APOSOR958338-1GEA
249
EUR
APOSOR958338-1G
APOSOR958338-250MG
cis-3-Carbomethoxycyclopentane-1-carboxylic acid 95%
cis-3-Carbomethoxycyclopentane-1-carboxylic acid
Formula:
C₈H₁₂O₄ MW: 172.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01311176 CAS Number: 96382-85-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...