5-Bromo[1,2,4]triazolo[4,3-a]pyridine 95%

Supplier: Apollo Scientific

Synonyms: 5-Bromo-[1,2,4]triazolo[4,3-a]pyridine

OR959510-1G OR959510-250MG
APOSOR959510-1GEA 267 EUR
APOSOR959510-1G APOSOR959510-250MG
5-Bromo[1,2,4]triazolo[4,3-a]pyridine 95%
5-Bromo[1,2,4]triazolo[4,3-a]pyridine
Formula: C₆H₄BrN₃
MW: 198,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11043995
CAS Number: 1172085-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR