Cyclohexyl-2-pyridyl ketone 95%

Supplier: Apollo Scientific

OR959516-1G OR959516-250MG
APOSOR959516-1GEA 502 EUR
APOSOR959516-1G APOSOR959516-250MG
Cyclohexyl-2-pyridyl ketone 95%
Cyclohexyl-2-pyridyl ketone
Formula: C₁₂H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD00778683
CAS Number: 6602-64-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR