2-Valerylpyridine 97%
Supplier: Apollo Scientific
OR959517-1G
OR959517-5G
APOSOR959517-1GEA
174
EUR
APOSOR959517-1G
APOSOR959517-5G
2-Valerylpyridine 97%
2-Valerylpyridine
Formula:
C₁₀H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD07699185 CAS Number: 7137-97-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...