5-Methylpyrimidin-2-ol hydrochloride
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Methylpyrimidin-2-ol
OR959838-1G
OR959838-5G
APOSOR959838-1GEA
33.5
EUR
APOSOR959838-1G
APOSOR959838-5G
5-Methylpyrimidin-2-ol hydrochloride
5-Methylpyrimidin-2-ol hydrochloride
Formula:
C₅H₇ClN₂O MW: 146,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20441449 CAS Number: 41398-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...