1-Ethyl-1H-pyrazole-3-carbaldehyde

Supplier: Apollo Scientific
Warning

OR959877-1G OR959877-5G OR959877-25G
APOSOR959877-1GEA 50.9 EUR
APOSOR959877-1G APOSOR959877-5G APOSOR959877-25G
1-Ethyl-1H-pyrazole-3-carbaldehyde
1-Ethyl-1H-pyrazole-3-carbaldehyde
Formula: C₆H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD03419802
CAS Number: 942319-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR