5-Bromo-2-iodobenzonitrile
Supplier: Apollo Scientific
|
Warning
|
OR9599-25G
OR9599-5G
APOSOR9599-25GEA
321
EUR
APOSOR9599-25G
APOSOR9599-5G
5-Bromo-2-iodobenzonitrile
5-Bromo-2-iodobenzonitrile
Formula:
C₇H₃BrIN MW: 307,92 g/mol Melting Pt: 120…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672950 CAS Number: 121554-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...