C-(3-Benzyl[1,2,4]oxadiazol-5-yl)methylamine 97%
Supplier: Apollo Scientific
OR960230-1G
OR960230-250MG
APOSOR960230-1GEA
377
EUR
APOSOR960230-1G
APOSOR960230-250MG
C-(3-Benzyl[1,2,4]oxadiazol-5-yl)methylamine 97%
C-(3-Benzyl[1,2,4]oxadiazol-5-yl)methylamine
Formula:
C₁₀H₁₁N₃O MW: 189,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07801111 CAS Number: 876715-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...