4-Bromofuran-2-one 95%

Supplier: Apollo Scientific

OR960467-100MG OR960467-1G OR960467-250MG
APOSOR960467-100MGEA 147 EUR
APOSOR960467-100MG APOSOR960467-1G APOSOR960467-250MG
4-Bromofuran-2-one 95%
4-Bromofuran-2-one
Formula: C₄H₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11109368
CAS Number: 56634-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR