3-Bromo-4H-thieno[3,2-b]pyrrole-5-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

OR960480-1G OR960480-250MG OR960480-5G
APOSOR960480-1GEA 183 EUR
APOSOR960480-1G APOSOR960480-250MG APOSOR960480-5G
3-Bromo-4H-thieno[3,2-b]pyrrole-5-carboxylic acid 95%
3-Bromo-4H-thieno[3,2-b]pyrrole-5-carboxylic acid
Formula: C7H4BrNO2S
Storage Temperature: Ambient
CAS Number: 332099-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR