Boc-D-Cha-OH 97%

Supplier: Apollo Scientific

Synonyms: Boc-β-cyclohexyl-D-Ala-OH

OR960780-1G OR960780-25G OR960780-5G
APOSOR960780-1GEA 52.4 EUR
APOSOR960780-1G APOSOR960780-25G APOSOR960780-5G
Boc-D-Cha-OH 97%
Boc-D-Cha-OH
Formula: C₁₄H₂₅NO₄
MW: 271,357 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133627
CAS Number: 127095-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR