4-Methyl-1H-indole-3-carbaldehyde 95%

Supplier: Apollo Scientific

Synonyms: 3-Formyl-4-methylindole

OR960850-1G OR960850-5G
APOSOR960850-1GEA 78.3 EUR
APOSOR960850-1G APOSOR960850-5G
4-Methyl-1H-indole-3-carbaldehyde 95%
4-Methyl-1H-indole-3-carbaldehyde
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049345
CAS Number: 4771-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR