2-Chloro-4-phenylpyridine 98%
Supplier: Apollo Scientific
OR961595-10G
OR961595-1G
OR961595-5G
APOSOR961595-10GEA
528
EUR
APOSOR961595-10G
APOSOR961595-1G
APOSOR961595-5G
2-Chloro-4-phenylpyridine 98%
2-Chloro-4-phenylpyridine
Formula:
C₁₁H₈ClN MW: 189.6400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04114163 CAS Number: 42260-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...