1-Cyclohexene-1-carboxylic acid 95%

Supplier: Apollo Scientific

Synonyms: Cyclohex-1-enecarboxylic acid

OR961865-25G
APOSOR961865-25GEA 278 EUR
APOSOR961865-25G
1-Cyclohexene-1-carboxylic acid 95%
1-Cyclohexene-1-carboxylic acid
Formula: C₇H₁₀O₂
MW: 126,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001545
CAS Number: 636-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR