2-Ethynylanisole 98%
Supplier: Apollo Scientific
Synonyms:
2'-Methoxyphenyl acetylene, 1-Ethynyl-2-methoxybenzene
OR961886-25G
OR961886-5G
APOSOR961886-25GEA
250
EUR
APOSOR961886-25G
APOSOR961886-5G
2-Ethynylanisole 98%
2-Ethynylanisole
Formula:
C₉H₈O MW: 132,1600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03839989 CAS Number: 767-91-9 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...