Allyl-4-oxopiperidine-1-carboxylate 95%
Supplier: Apollo Scientific
Synonyms:
1-N-Alloc-4-piperidone
OR962024-1G
APOSOR962024-1GEA
328
EUR
APOSOR962024-1G
Allyl-4-oxopiperidine-1-carboxylate 95%
Allyl-4-oxopiperidine-1-carboxylate
Formula:
C₉H₁₃NO₃ MW: 183,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02183578 CAS Number: 306296-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...