Methyl 5-bromo-1H-indole-3-carboxylate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 5-bromo-1H-indole-3-carboxylate

OR962104-25G OR962104-5G
APOSOR962104-25GEA 288 EUR
APOSOR962104-25G APOSOR962104-5G
Methyl 5-bromo-1H-indole-3-carboxylate 98%
Methyl-5-bromo-1H-indole-3-carboxylate
Formula: C₁₀H₈BrNO₂
MW: 254.0800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203728
CAS Number: 773873-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR